Learn More
Aviva Systems Biology Corp RNF10 Antibody - N-terminal region (ARP34336_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34336_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF10. Alternate names: KIAA0262, MGC126758, MGC126764, RIE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55346