Learn More
Aviva Systems Biology Corp RNF113A Antibody - middle region (ARP38892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF113A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF113A. Alternate names: RNF113, ZNF183, Cwc24ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-56837