Learn More
Aviva Systems Biology Corp RNF113B Antibody - C-terminal region (ARP40028_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40028_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF113B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNF113B. Alternate names: RNF161, ZNF183L1, bA10G5.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57270