Learn More
Aviva Systems Biology Corp RNF113B Antibody - middle region (ARP34808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF113B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF113B. Alternate names: MGC26599, RNF161, ZNF183L1, bA10G5.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55572