Learn More
Aviva Systems Biology Corp RNF11L ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP43148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rnf11l. It was validated on Western Blot. . Alternate names: Rnf11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-058