Learn More
Aviva Systems Biology Corp RNF12 Antibody - C-terminal region (ARP32151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF12. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RNF12. Alternate names: RNF12, NY-REN-43ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54466