Learn More
Aviva Systems Biology Corp RNF12 Antibody - N-terminal region (ARP32150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF12. Alternate names: MGC15161, NY-REN-43, RLIM, RNF12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54465