Learn More
Aviva Systems Biology Corp Rnf122 Antibody - middle region (ARP43320_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43320_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rnf122. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110063C11RikApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-066