Learn More
Aviva Systems Biology Corp RNF125 Antibody - middle region (ARP43234_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43234_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF125. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF125. Alternate names: FLJ20456, MGC21737, TRAC1, TRAC-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62831