Learn More
Aviva Systems Biology Corp RNF126 Antibody - N-terminal region (ARP43235_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43235_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF126. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF126. Alternate names: FLJ20552, MGC1022, MGC14317ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58468