Learn More
Aviva Systems Biology Corp RNF128 Antibody - C-terminal region (ARP43311_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43311_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF128. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNF128. Alternate names: FLJ23516, GRAILApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53362