Learn More
Aviva Systems Biology Corp RNF128 Antibody - C-terminal region (ARP59127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF128. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RNF128. Alternate names: FLJ23516, GRAILApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-588