Learn More
Aviva Systems Biology Corp RNF135 Antibody - middle region (ARP34886_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34886_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF135. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF135. Alternate names: L13, MGC13061, MMFD, REUL, RipletApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-55600