Learn More
Aviva Systems Biology Corp RNF138 Antibody - C-terminal region (ARP33005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF138. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNF138. Alternate names: HSD-4, MGC8758, NARF, STRIN, hNARFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54849