Learn More
Aviva Systems Biology Corp RNF14 Antibody - C-terminal region (ARP38448_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38448_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF14. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RNF14. Alternate names: ARA54, HFB30, TRIAD2, HRIHFB2038ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Rabbit, Rat
Catalog No. 50-014-56660