Learn More
Aviva Systems Biology Corp RNF14 Antibody - N-terminal region (ARP38447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF14. Alternate names: ARA54, FLJ26004, HFB30, HRIHFB2038, TRIAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56659