Learn More
Aviva Systems Biology Corp RNF141 Antibody - middle region (ARP34554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF141. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RNF141. Alternate names: ZFP26, ZNF230ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55447