Learn More
Aviva Systems Biology Corp RNF144B Antibody - middle region (ARP43479_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43479_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF144B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF144B. Alternate names: IBRDC2, KIAA0161, MGC71786, bA528A10.3, p53RFP, PIR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58544