Learn More
Aviva Systems Biology Corp RNF148 Antibody - C-terminal region (ARP43494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF148. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNF148. Alternate names: MGC35222ApplicationWB. Species reactivity: Human, Rat, Yeast
Catalog No. 50-014-53365