Learn More
Aviva Systems Biology Corp RNF165 Antibody - N-terminal region (ARP43418_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43418_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF165. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF165. Alternate names: ARKL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58525