Learn More
Aviva Systems Biology Corp RNF167 Antibody - middle region (ARP43199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF167. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RNF167. Alternate names: 5730408C10Rik, DKFZP566H073, LP2254, RING105ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-062