Learn More
Aviva Systems Biology Corp RNF168 Antibody - C-terminal region (ARP43433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF168. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RNF168. Alternate names: FLJ35794ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58531