Learn More
Aviva Systems Biology Corp RNF170 Antibody - middle region (ARP43337_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43337_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF170. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF170. Alternate names: DKFZP564A022, FLJ38306ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58502