Learn More
Aviva Systems Biology Corp RNF182 Antibody - N-terminal region (ARP43438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF182. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF182. Alternate names: FLJ40772, MGC33993ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58534