Learn More
Aviva Systems Biology Corp Rnf183 Antibody - N-terminal region (ARP43404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rnf183. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 5830442J12RikApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-070