Learn More
Aviva Systems Biology Corp RNF186 Antibody - N-terminal region (ARP43262_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43262_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF186. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF186. Alternate names: FLJ20225, RP11-91K11.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58477