Learn More
Aviva Systems Biology Corp RNF19A Antibody - N-terminal region (ARP43197_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43197_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF19A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF19A. Alternate names: DKFZp566B1346, DORFIN, RNF19ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53360