Learn More
Aviva Systems Biology Corp RNF2 Antibody - middle region (ARP34290_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34290_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RNF2. Alternate names: BAP1, DING, BAP-1, HIPI3, RING2, RING1BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55327