Learn More
Aviva Systems Biology Corp RNF20 Antibody - N-terminal region (ARP43264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF20. Alternate names: BRE1A, FLJ11189, FLJ20382, KIAA2779, MGC129667, MGC129668, BRE1, hBRE1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58479