Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci RNF20 Peptide
SDP

Catalog No. 16597127
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-597-127 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-597-127 Supplier ProSci Supplier No. 7357P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

RNF20 peptide is used for blocking the activity of RNF20 antibody (catalog number 7357). It usually blocks the antibody activity completely in Western blot by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 56254
Species Human
Accession Number NP_062538
Common Name ring finger protein 20, E3 ubiquitin protein ligase
Concentration 200 μg/mL
Content And Storage Store RNF20 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol RNF20
Quantity 0.05 mg
Sequence 19 amino acid peptide near the amino terminus of human RNF20.
Synonym BRE1, BRE1A, hBRE1, E3 ubiquitin-protein ligase BRE1A, RING finger protein 20, BRE1-A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.