Learn More
Aviva Systems Biology Corp RNF212 Antibody - middle region (ARP43487_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF212. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF212. Alternate names: FLJ38841, MGC120227, MGC120228, ZHP3ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit
Catalog No. 50-014-58549