Learn More
Aviva Systems Biology Corp RNF31 Antibody - middle region (ARP43241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF31. Alternate names: FLJ10111, FLJ23501, MGC19975, ZIBRA, HOIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58470