Learn More
Aviva Systems Biology Corp RNF39 Antibody - C-terminal region (ARP43447_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43447_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RNF39. Alternate names: HZF, HZFW, LIRFApplicationWB. Species reactivity: Human
Catalog No. 50-014-58537