Learn More
Aviva Systems Biology Corp RNF39 Antibody - N-terminal region (ARP43332_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43332_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF39. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RNF39. Alternate names: HZF, HZFW, HZFw1, LIRFApplicationIHC, WB. Species reactivity: Horse, Human
Catalog No. 50-014-58500