Learn More
Aviva Systems Biology Corp RNF40 Antibody - C-terminal region (ARP43165_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43165_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF40. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RNF40. Alternate names: BRE1B, DKFZp686K191, KIAA0661, MGC13051, RBP95, STARINGApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53359