Learn More
Aviva Systems Biology Corp RNF41 Antibody - middle region (ARP34694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF41. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNF41. Alternate names: MGC45228, NRDP1, SBBI03, FLRFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55516