Learn More
Aviva Systems Biology Corp RNF44 Antibody - N-terminal region (ARP43171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF44. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF44. Alternate names: KIAA1100ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58455