Learn More
Medchemexpress LLC Rnf5 inhibitor inh-02 | 324579-65-9 | 99.9% | 384.50 g·mol-1 | C23H20N4S | 25MG
SDP
Catalog No. 5000237276
Encompass_Preferred
RNF5 inhibitor inh-02 is a small-molecule inhibitor of the E3 ubiquitin ligase RNF5 used in research to modulate protein ubiquitylation, autophagy, and to rescue F508del-CFTR function in cellular models (reported cellular EC50 ≈ 2.2-2.6 μM). For research use only; not for human use.
Catalog No. 50-002-37276 Supplier Medchemexpress LLC Supplier No. HY12396725MG
RNF5 inhibitor inh-02 is a small-molecule inhibitor of the E3 ubiquitin ligase RNF5 used in research to modulate protein ubiquitylation, autophagy, and to rescue F508del-CFTR function in cellular models (reported cellular EC50 ≈ 2.2-2.6 μM). For research use only; not for human use.