Learn More
Medchemexpress LLC RNF5 inhibitor inh-02 | 324579-65-9 | 99.9% | 384.50 g/mol | C23H20N4S | 100MG
SDP
Catalog No. 5000238147
Encompass_Preferred
RNF5 inhibitor inh-02 is a research-grade small-molecule inhibitor of the E3 ubiquitin ligase RNF5, used in cellular and biochemical studies to probe RNF5-dependent ubiquitination and reported to rescue F508del-CFTR function in experimental models. The substance is supplied as a solid with high reported purity and is available in multiple pack sizes and as ready-to-use DMSO solutions.
Catalog No. 50-002-38147 Supplier Medchemexpress LLC Supplier No. HY123967100MG
RNF5 inhibitor inh-02 is a research-grade small-molecule inhibitor of the E3 ubiquitin ligase RNF5, used in cellular and biochemical studies to probe RNF5-dependent ubiquitination and reported to rescue F508del-CFTR function in experimental models. The substance is supplied as a solid with high reported purity and is available in multiple pack sizes and as ready-to-use DMSO solutions.