Learn More
Aviva Systems Biology Corp RNF6 Antibody - N-terminal region (ARP37397_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37397_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of mouse RNF6. Alternate names: AA537053, 1200013I08Rik, 5730419H05RikApplicationIHC, WB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56363