Learn More
Aviva Systems Biology Corp RNF6 Antibody - N-terminal region (ARP43064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF6. Alternate names: DKFZp686P0776ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-58413