Learn More
Cayman Chemical RnKLODF EXrAClular DomaI 100ug
SDP
Catalog No. 502942244
Encompass_Preferred
Source: Active recombinant N-terminal rabbit IgG Fc-tagged human RANKL/ODF extracellular domain expressed in HEK293 cells • Amino acids: 64-245 • MW: 46.18 kDa
Catalog No. 50-294-2244 Supplier Cayman Chemical Supplier No. 40209100 UG
Source: Active recombinant N-terminal rabbit IgG Fc-tagged human RANKL/ODF extracellular domain expressed in HEK293 cells • Amino acids: 64-245 • MW: 46.18 kDa