Learn More
Aviva Systems Biology Corp Rnls Antibody - middle region (ARP59612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rnls. It was validated on Western Blot. . Alternate names: 6530404N21Rik, AI452315, AW060440, C10orf59ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-757