Learn More
Aviva Systems Biology Corp RNMT Antibody - N-terminal region (ARP40494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNMT. Alternate names: DKFZp686H1252, KIAA0398, MET, RG7MT1, hCMT1cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-57485