Learn More
Medchemexpress LLC HY-12824 5mg Medchemexpress, RNPA1000 CAS:359600-10-5 Purity:>98%
SDP
Catalog No. 502081159
Encompass_Preferred
Medchemexpress, HY-12824 5mg RNPA1000 CAS:359600-10-5 RNPA1000, an antibiotic, is a potent RnpA inhibitor and inhibits RnpA-mediated cellular RNA degradation. RNPA1000 inhibits tRNA maturation with an IC50 of 175 μM. RNPA1000 displays broad-spectrum antimicrobial activities and inhibits staphylococcal and all Gram-positive bacterial pathogens activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1159 Supplier Medchemexpress LLC Supplier No. HY128245MG
Medchemexpress, HY-12824 5mg RNPA1000 CAS:359600-10-5 RNPA1000, an antibiotic, is a potent RnpA inhibitor and inhibits RnpA-mediated cellular RNA degradation. RNPA1000 inhibits tRNA maturation with an IC50 of 175 μM. RNPA1000 displays broad-spectrum antimicrobial activities and inhibits staphylococcal and all Gram-positive bacterial pathogens activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.