Learn More
Aviva Systems Biology Corp RNPC3 Antibody - middle region (ARP40868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RNPC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RNPC3. Alternate names: FLJ20008, FLJ25070, KIAA183, RBM40, RNPApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-57645