Learn More
Medchemexpress LLC RNS peptide | 901117-08-6 | 99.2% | 2498.83 | 5 MG
SDP
Catalog No. 5000424061
Encompass_Preferred
RNS peptide is a randomly scrambled control peptide that can be used as a negative control for MANS peptide. It is supplied as a white to off-white solid with high purity.
  • Can be used as a negative control
  • White to off-white solid appearance
  • Sealed storage, away from moisture and light
  • Powder storage: -80°C for 2 years or -20°C for 1 year
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-24061 Supplier Medchemexpress LLC Supplier No. HYP107765MG
RNS peptide is a randomly scrambled control peptide that can be used as a negative control for MANS peptide. It is supplied as a white to off-white solid with high purity.
  • Can be used as a negative control
  • White to off-white solid appearance
  • Sealed storage, away from moisture and light
  • Powder storage: -80°C for 2 years or -20°C for 1 year
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL