Learn More
Cayman Chemical Ro 046790hydroCLOrID hyd 10mg
SDP
Catalog No. 502946778
Encompass_Preferred
A 5-HT6 receptor antagonist (Ki = 44.7 nM); selective for 5-HT6 receptors over a panel of 23 additional receptors at 10 µM; inhibits 5-HT-induced cAMP production in HeLa cells expressing human 5-HT6 (pA2 = 6.75); induces stretching and yawning behavior in rats at 10 and 30 mg/kg; reverses scopolamine-induced decreases in novel object discrimination in rats; attenuates cue-induced cocaine-seeking behavior in rats at 3, 10, and 30 mg/kg
Catalog No. 50-294-6778 Supplier Cayman Chemical Supplier No. 3447610 MG
A 5-HT6 receptor antagonist (Ki = 44.7 nM); selective for 5-HT6 receptors over a panel of 23 additional receptors at 10 µM; inhibits 5-HT-induced cAMP production in HeLa cells expressing human 5-HT6 (pA2 = 6.75); induces stretching and yawning behavior in rats at 10 and 30 mg/kg; reverses scopolamine-induced decreases in novel object discrimination in rats; attenuates cue-induced cocaine-seeking behavior in rats at 3, 10, and 30 mg/kg