Learn More
TARGETMOL CHEMICALS INC RO 08-2750 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167665MG
Encompass_Preferred
Also available in 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro 08-2750 is a non-peptide and reversible nerve growth factor (NGF) inhibitor which binds to NGF and with an IC50 of ~ 1 µM. Ro 08-2750 is a selective MSI RNA-binding activity inhibitor with an IC50 of 2.7 uM. Ro 08-2750 inhibits NGF binding to p75NTR selectively over TRKA. purity: 98%
Catalog No. 50-258-1156