Learn More
Abcam Ro 08-2750, 10MG
Catalog No. 502681667
Encompass_Preferred
MW 270.24 Da, Purity =98%. An antagonist of nerve growth factor (NGF) with binding ratio of 1:1, IC₅₀ = 1 μM. Selectively inhibits binding to p75NTR over TRKA147 at low concentrations. However, it can impair NGF binding to TrkA at higher concentrations (more than 5 mM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-1667 Supplier Abcam Supplier No. AB28243610MG
MW 270.24 Da, Purity =98%. An antagonist of nerve growth factor (NGF) with binding ratio of 1:1, IC₅₀ = 1 μM. Selectively inhibits binding to p75NTR over TRKA147 at low concentrations. However, it can impair NGF binding to TrkA at higher concentrations (more than 5 mM).
The product is subject to the following: Abcam Restricted Use Statement